4,7-Bis(5-bromo-4-hexylthiophen-2-yl)benzo[c][1,2,5]thiadiazole 98%
Supplier: Ambeed
|
Warning
|
A187511-100MG
A187511-1G
A187511-250MG
77236-734EA
52.53
USD
77236-734
77236-736
77236-738
4,7-Bis(5-bromo-4-hexylthiophen-2-yl)benzo[c][1,2,5]thiadiazole 98%
4,7-Bis(5-bromo-4-hexylthiophen-2-yl)benzo[c][1,2,5]thiadiazole
Precaución: for Research Use Only
Formula:
C₂₆H₃₀Br₂N₂S₃ MW: 626.53 g/mol |
MDL Number:
MFCD30186156 CAS Number: 444579-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...