2-(3,5-Dichlorophenyl)propan-2-amine 97%

Supplier: Ambeed
Warning

A187452-10G A187452-1G A187452-250MG A187452-5G
77248-456EA 373.89 USD
77248-456 77248-458 77248-460 77248-462
2-(3,5-Dichlorophenyl)propan-2-amine 97%
2-(3,5-Dichlorophenyl)propan-2-amine

Precaución: for Research Use Only

Formula: C₉H₁₁Cl₂N
MW: 204.10 g/mol
MDL Number: MFCD11846158
CAS Number: 129960-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR