3-Fluoropyridine-2-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-fluoropicolinate

A187404-1G A187404-250MG A187404-5G
77260-430EA 81.37 USD
77260-430 77260-432 77260-434
3-Fluoropyridine-2-carboxylic acid tert-butyl ester 95%
3-Fluoropyridine-2-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₂
MW: 197.206 g/mol
MDL Number: MFCD29761614
CAS Number: 1934827-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR