2,2-Difluoroethyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

Synonyms: 2,2-Difluoroethyltrifluoromethanesulphonate

A187403-1G A187403-25G A187403-5G
77248-454EA 6.18 USD
77248-454 77550-200 77567-744
2,2-Difluoroethyl trifluoromethanesulfonate 95%
2,2-Difluoroethyl trifluoromethanesulfonate

Precaución: For research use only.

Formula: C₃H₃F₅O₃S
MW: 214.11 g/mol
MDL Number: MFCD02093342
CAS Number: 74427-22-8
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR