Fmoc-3-amino-4-methoxybenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybenzoic acid

A187317-1G A187317-250MG A187317-5G
77236-718EA 31.93 USD
77236-718 77236-720 77236-722
Fmoc-3-amino-4-methoxybenzoic acid 97%
Fmoc-3-amino-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₂₃H₁₉NO₅
MW: 389.41 g/mol
MDL Number: MFCD02682216
CAS Number: 256935-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR