3-Pyrrolidin-1-ylpropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Pyrrolidin-1-yl)propanoic acid

A186768-10G A186768-1G A186768-250MG A186768-5G
77287-416EA 247 USD
77287-416 77287-418 77287-420 77287-422
3-Pyrrolidin-1-ylpropanoic acid 95%
3-Pyrrolidin-1-ylpropanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD04114076
CAS Number: 76234-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR