2,4-Dichloro-6-methyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

A186705-1G A186705-250MG A186705-5G
77248-434EA 13 USD
77248-434 77248-436 77248-438
2,4-Dichloro-6-methyl-3-nitropyridine 98%
2,4-Dichloro-6-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
MDL Number: MFCD00218548
CAS Number: 63897-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR