PF-9366 98%

Supplier: Ambeed
Warning

Synonyms: 2-(7-Chloro-5-phenyl-[1,2,4]triazolo[4,3-a]quinolin-1-yl)-N,N-dimethylethan-1-amine

A186389-10MG A186389-1MG A186389-25MG A186389-5MG A186389-100MG A186389-250MG A186389-50MG
77260-418EA 50 USD
77260-418 77260-420 77260-422 77260-424 77748-209 77748-210 77748-211
PF-9366 98%
PF-9366

Precaución: for Research Use Only

Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
MDL Number: MFCD31630724
CAS Number: 72882-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR