Chloroacetamidine hydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: 2-Chloroacetimidamide hydrochloride, 2-Chloroacetamidine hydrochloride

A186338-10G A186338-1G A186338-250MG A186338-5G A186338-25G A186338-100G
77260-408EA 42 USD
77260-408 77260-410 77260-412 77260-414 77615-576 77748-208
Chloroacetamidine hydrochloride 97%
Chloroacetamidine hydrochloride

Precaución: for Research Use Only

Formula: C₂H₆Cl₂N₂
MDL Number: MFCD00053013
CAS Number: 10300-69-3
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR