3-Bromo-5-methylbenzenesulfonyl chloride 97%

Supplier: Ambeed
Danger

A186284-10G A186284-1G A186284-250MG A186284-5G
77274-306EA 313 USD
77274-306 77274-308 77274-310 77274-312
3-Bromo-5-methylbenzenesulfonyl chloride 97%
3-Bromo-5-methylbenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrClO₂S
MDL Number: MFCD07781544
CAS Number: 885520-33-2
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR