HMN-176 98%

Supplier: Ambeed
Warning

Synonyms: (E)-4-(2-((4-Methoxyphenyl)sulphonamido)styryl)pyridine 1-oxide

A186241-10MG A186241-1MG A186241-25MG A186241-50MG A186241-5MG
77260-398EA 109.18 USD
77260-398 77260-400 77260-402 77260-404 77260-406
HMN-176 98%
HMN-176

Precaución: for Research Use Only

Formula: C₂₀H₁₈N₂O₄S
MW: 382.43 g/mol
MDL Number: MFCD00945108
CAS Number: 173529-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR