3-(Trifluoromethyl)-1H-indole 95%

Supplier: Ambeed
Warning

A186063-1G A186063-250MG
77287-408EA 1061.93 USD
77287-408 77593-198
3-(Trifluoromethyl)-1H-indole 95%
3-(Trifluoromethyl)-1H-indole

Precaución: for Research Use Only

Formula: C₉H₆F₃N
MW: 185.15 g/mol
MDL Number: MFCD09032290
CAS Number: 51310-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR