(4-(Morpholinomethyl)phenyl)methanamine 95%

Supplier: Ambeed
Warning

A185993-1G A185993-250MG
77274-286EA 41 USD
77274-286 77274-288
(4-(Morpholinomethyl)phenyl)methanamine 95%
(4-(Morpholinomethyl)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD05664410
CAS Number: 91271-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR