N-(4-(4-(2-Methoxyphenyl)piperazin-1-yl)butyl)-2-naphthamide hydrochloride 98%

Supplier: Ambeed
Warning

A185949-10MG A185949-1MG A185949-25MG A185949-50MG A185949-5MG
77274-276EA 87 USD
77274-276 77274-278 77274-280 77274-282 77274-284
N-(4-(4-(2-Methoxyphenyl)piperazin-1-yl)butyl)-2-naphthamide hydrochloride 98%
N-(4-(4-(2-Methoxyphenyl)piperazin-1-yl)butyl)-2-naphthamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₆H₃₂ClN₃O₂
MDL Number: MFCD04039996
CAS Number: 314776-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR