5'-Chloro-2'-methoxyacetophenone 95%

Supplier: Ambeed
Warning

A185891-1G A185891-250MG A185891-25G A185891-5G
77274-268EA 6 USD
77274-268 77274-270 77274-272 77274-274
5'-Chloro-2'-methoxyacetophenone 95%
5'-Chloro-2'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD00174339
CAS Number: 6342-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR