5-Hydroxy-4-methyl-2(5H)furanone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Hydroxy-4-methylfuran-2(5H)-one
A185843-100G
A185843-10G
A185843-25G
77287-396EA
104.03
USD
77287-396
77287-398
77287-400
5-Hydroxy-4-methyl-2(5H)furanone 97%
5-Hydroxy-4-methyl-2(5H)furanone
Precaución: for Research Use Only
Formula:
C₅H₆O₃ MW: 114.099 g/mol |
MDL Number:
MFCD07782086 CAS Number: 40834-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...