2-Cyclopropyl-3-nitro-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 3-Cyclopropyl-4-nitro-1H-pyrazole

A185808-100MG A185808-1G A185808-250MG
77248-416EA 22.66 USD
77248-416 77248-418 77248-420
2-Cyclopropyl-3-nitro-1H-pyrazole 97%
2-Cyclopropyl-3-nitro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.139 g/mol
MDL Number: MFCD16249530
CAS Number: 1249581-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR