1-(2-Morpholinoethyl)-3-(4-(3-(6-morpholinopyridin-2-yl)acryloyl)phenyl)urea hydrochloride 98%

Supplier: Ambeed
Warning

A185676-10MG A185676-25MG A185676-50MG A185676-5MG
77260-362EA 102 USD
77260-362 77260-364 77260-366 77260-368 77260-370
1-(2-Morpholinoethyl)-3-(4-(3-(6-morpholinopyridin-2-yl)acryloyl)phenyl)urea hydrochloride 98%
1-(2-Morpholinoethyl)-3-(4-(3-(6-morpholinopyridin-2-yl)acryloyl)phenyl)urea hydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₃₂ClN₅O₄
MDL Number: MFCD31539844
CAS Number: 1333327-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR