4-Benzyloxyindole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A185639-100MG A185639-1G A185639-250MG
77274-262EA 42.23 USD
77274-262 77274-264 77274-266
4-Benzyloxyindole-3-carbaldehyde 98%
4-Benzyloxyindole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₂
MW: 251.28 g/mol
MDL Number: MFCD00152002
CAS Number: 7042-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR