2-Amino-3,5-diiodopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Diiodo-2-pyridylamine, 3,5-Diiodo-2-Pyridinamine, (3,5-Diiodo-2-Pyridyl)Amine, 3,5-Diiodopyridin-2-amine, 2-amino-3,5-diiodopyridine

A185489-1G A185489-250MG A185489-25G A185489-5G
77248-404EA 12.36 USD
77248-404 77248-406 77248-408 77248-410
2-Amino-3,5-diiodopyridine 98%
2-Amino-3,5-diiodopyridine

Precaución: for Research Use Only

Formula: C₅H₄I₂N₂
MW: 345.91 g/mol
MDL Number: MFCD02663852
CAS Number: 23597-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR