(S)-(+)-2-Phenylglycine methyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 2-amino-2-phenylacetate hydrochloride, H-Phg-OMe·HCl, L-(+)-2-Phenylglycine methyl ester hydrochloride

A185347-100G A185347-10G A185347-25G A185347-500G A185347-5G
77248-390EA 43 USD
77248-390 77248-392 77248-394 77248-396 77248-398
(S)-(+)-2-Phenylglycine methyl ester hydrochloride 98%
(S)-(+)-2-Phenylglycine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂
MW: 201.65 g/mol
MDL Number: MFCD00077158
CAS Number: 15028-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR