4-Aminopyridine-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Aminopicolinonitrile

A185179-100MG A185179-250MG A185179-1G
77287-380EA 18 USD
77287-380 77287-382 77748-203
4-Aminopyridine-2-carbonitrile 97%
4-Aminopyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₅N₃
MDL Number: MFCD07374351
CAS Number: 98139-15-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR