3-Methoxycarbonyl-5-nitrobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, 3-Methoxycarbonyl-5-nitrophenylboronic acid, 3-(Methoxycarbonyl)-5-nitrophenylboronic acid, (3-(Methoxycarbonyl)-5-nitrophenyl)boronic acid

A185035-100G A185035-10G A185035-1G A185035-25G A185035-5G
77236-686EA 351.01 USD
77236-686 77236-688 77236-690 77236-692 77236-694
3-Methoxycarbonyl-5-nitrobenzeneboronic acid 98%
3-Methoxycarbonyl-5-nitrobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₆
MW: 224.96 g/mol
MDL Number: MFCD02179469
CAS Number: 117342-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR