1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-benzo[d][1,2,3]triazole-5-carbaldehyde

A184946-100MG A184946-10G A184946-1G A184946-5G
77236-678EA 33 USD
77236-678 77236-680 77236-682 77236-684
1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde 98%
1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₇N₃O
MDL Number: MFCD03407337
CAS Number: 499770-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR