1-Chloroethylethyl carbonate 98%

Supplier: Ambeed
Warning

A184670-100G A184670-5G A184670-25G
77236-672EA 39 USD
77236-672 77236-674 77593-184
1-Chloroethylethyl carbonate 98%
1-Chloroethylethyl carbonate

Precaución: for Research Use Only

Formula: C₅H₉ClO₃
MDL Number: MFCD00040523
CAS Number: 50893-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR