6-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A184603-100MG A184603-1G A184603-250MG A184603-5G
77236-656EA 36.05 USD
77236-656 77236-658 77236-660 77236-662
6-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one 97%
6-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
MDL Number: MFCD21879587
CAS Number: 724422-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR