2-Methyl-5-nitronicotinic acid 97%

Supplier: Ambeed
Warning

A184537-100MG A184537-1G A184537-250MG A184537-5G
77274-232EA 16 USD
77274-232 77274-234 77274-236 77274-238
2-Methyl-5-nitronicotinic acid 97%
2-Methyl-5-nitronicotinic acid
Formula: C₇H₆N₂O₄
MDL Number: MFCD07339171
CAS Number: 59290-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR