3',4'-(Methylenedioxy)butyrophenone 98%

Supplier: Ambeed
Warning

A184479-100G A184479-1G A184479-25G A184479-5G
77274-224EA 333.99 USD
77274-224 77274-226 77274-228 77274-230
3',4'-(Methylenedioxy)butyrophenone 98%
3',4'-(Methylenedioxy)butyrophenone

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MDL Number: MFCD01075043
CAS Number: 63740-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR