3-Isopropylaniline 97%

Supplier: Ambeed
Warning

A184405-10G A184405-500G A184405-5G
77274-218EA 12 USD
77274-218 77274-220 77274-222
3-Isopropylaniline 97%
3-Isopropylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃N
MDL Number: MFCD00092918
CAS Number: 5369-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR