4-Phenyl-azetidin-2-one 98%

Supplier: Ambeed
Warning

Synonyms: 4-Phenylazetidin-2-one

A184350-100MG A184350-250MG A184350-1G
77287-362EA 39.14 USD
77287-362 77287-364 77748-198
4-Phenyl-azetidin-2-one 98%
4-Phenyl-azetidin-2-one

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.174 g/mol
MDL Number: MFCD02179052
CAS Number: 5661-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR