3,4-Difluoro-N-methylaniline 97%

Supplier: Ambeed
Danger

A184167-100MG A184167-1G A184167-250MG A184167-5G
77260-346EA 23.69 USD
77260-346 77260-348 77260-350 77260-352
3,4-Difluoro-N-methylaniline 97%
3,4-Difluoro-N-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂N
MW: 143.13 g/mol
MDL Number: MFCD11152726
CAS Number: 138563-54-9
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR