5-Ethynylpyridin-3-amine 97%

Supplier: Ambeed
Warning

A184126-100MG A184126-1G A184126-250MG
77260-340EA 70.04 USD
77260-340 77260-342 77260-344
5-Ethynylpyridin-3-amine 97%
5-Ethynylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD18207401
CAS Number: 667932-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR