5-Fluoropyrimidin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-fluoropyrimidine

A184053-10G A184053-1G A184053-250MG A184053-25G A184053-5G
77274-202EA 95 USD
77274-202 77274-204 77274-206 77274-208 77274-210
5-Fluoropyrimidin-2-amine 98%
5-Fluoropyrimidin-2-amine

Precaución: for Research Use Only

Formula: C4H4FN3
MDL Number: MFCD11846500
CAS Number: 1683-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR