(2R,4R)-Ethyl 4-methyl-1-((S)-2-(3-methylquinoline-8-sulfonamido)-5-(3-nitroguanidino)pentanoyl)piperidine-2-carboxylate 95%

Supplier: Ambeed
Danger

A184011-100MG A184011-250MG
77274-198EA 19.57 USD
77274-198 77274-200
(2R,4R)-Ethyl 4-methyl-1-((S)-2-(3-methylquinoline-8-sulfonamido)-5-(3-nitroguanidino)pentanoyl)piperidine-2-carboxylate 95%
(2R,4R)-Ethyl 4-methyl-1-((S)-2-(3-methylquinoline-8-sulfonamido)-5-(3-nitroguanidino)pentanoyl)piperidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₂₅H₃₅N₇O₇S
MW: 577.65 g/mol
MDL Number: MFCD08458425
CAS Number: 74874-09-2
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR