3-Amino-3-(4-chlorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-3-(4-chlorophenyl)propanoic acid, 3-Amino-3-(4-chlorophenyl)propionic Acid, 3-Amino-3-(p-chlorophenyl)propionic acid

A183860-1G A183860-25G A183860-5G
77248-358EA 11.33 USD
77248-358 77248-360 77248-362
3-Amino-3-(4-chlorophenyl)propionic acid 97%
3-Amino-3-(4-chlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
MDL Number: MFCD00126370
CAS Number: 19947-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR