(S)-tert-Butyl 4-(6-amino-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl)-3-methylpiperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A183846-100MG A183846-1G A183846-250MG
77236-644EA 21.63 USD
77236-644 77236-646 77236-648
(S)-tert-Butyl 4-(6-amino-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl)-3-methylpiperazine-1-carboxylate 97%
(S)-tert-Butyl 4-(6-amino-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl)-3-methylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₇N₇O₂
MW: 361.44 g/mol
MDL Number: MFCD30475784
CAS Number: 1616415-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR