(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(4-chloro-3-(4-(((S)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate 98%

Supplier: Ambeed
Warning

A183839-1G A183839-5G
77236-642EA 8 USD
77236-642 77615-554
(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(4-chloro-3-(4-(((S)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate 98%
(2R,3R,4R,5S,6S)-2-(Acetoxymethyl)-6-(4-chloro-3-(4-(((S)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate

Precaución: for Research Use Only

Formula: C₃₁H₃₅ClO₁₁
MDL Number: MFCD28978546
CAS Number: 915095-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR