4-(2,2-Diethoxyethyl)morpholine 97%
Supplier: Ambeed
|
Warning
|
A183658-1G
A183658-250MG
A183658-25G
A183658-5G
77287-342EA
14.42
USD
77287-342
77287-344
77287-346
77287-348
4-(2,2-Diethoxyethyl)morpholine 97%
4-(2,2-Diethoxyethyl)morpholine
Precaución: for Research Use Only
Formula:
C₁₀H₂₁NO₃ MW: 203.27 g/mol |
MDL Number:
MFCD00034483 CAS Number: 3616-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...