7-Bromo-1-chloroisoquinoline 97%

Supplier: Ambeed
Warning

A183570-1G A183570-250MG A183570-25G A183570-5G
77260-306EA 17 USD
77260-306 77260-308 77260-310 77615-552
7-Bromo-1-chloroisoquinoline 97%
7-Bromo-1-chloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MDL Number: MFCD04039320
CAS Number: 215453-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR