(E)-3-(3-chlorophenyl)-N-(2-((1,1-dioxidotetrahydro-2H-thiopyran-4-yl)(methyl)amino)-2-oxoethyl)acrylamide 98%

Supplier: Ambeed
Warning

A183374-1MG A183374-50MG A183374-5MG
77236-636EA 22 USD
77236-636 77236-638 77236-640
(E)-3-(3-chlorophenyl)-N-(2-((1,1-dioxidotetrahydro-2H-thiopyran-4-yl)(methyl)amino)-2-oxoethyl)acrylamide 98%
(E)-3-(3-chlorophenyl)-N-(2-((1,1-dioxidotetrahydro-2H-thiopyran-4-yl)(methyl)amino)-2-oxoethyl)acrylamide

Precaución: for Research Use Only

Formula: C₁₇H₂₁ClN₂O₄S
MDL Number: MFCD30187519
CAS Number: 1550008-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR