Ixazomib citrate 98%

Supplier: Ambeed
Warning

A183051-10MG A183051-1MG A183051-2MG
77236-624EA 18 USD
77236-624 77236-626 77236-628
Ixazomib citrate 98%
Ixazomib citrate

Precaución: for Research Use Only

Formula: C₂₀H₂₃BCl₂N₂O₉
MDL Number: MFCD29767859
CAS Number: 1239908-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR