2-Ethyl-2-methyl-1,3-dioxolane 96%

Supplier: Ambeed
Danger

A182940-25G A182940-5G
77260-292EA 54.59 USD
77260-292 77260-294
2-Ethyl-2-methyl-1,3-dioxolane 96%
2-Ethyl-2-methyl-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
MDL Number: MFCD00014108
CAS Number: 126-39-6
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR