Ethyl 2-(tert-butyl)thiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A182930-100MG A182930-250MG
77236-620EA 182.31 USD
77236-620 77236-622
Ethyl 2-(tert-butyl)thiazole-5-carboxylate 97%
Ethyl 2-(tert-butyl)thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂S
MW: 213.29 g/mol
MDL Number: MFCD28137670
CAS Number: 1012881-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR