(S,S)-Ts-DENEB® 98%

Supplier: Ambeed
Danger

Synonyms: Chloro[(S,S)-N-[2-(4-methylbenzyloxy)ethyl]-N'-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), N-[(1S,2S)-1,2-Diphenyl-2-(2-(4-methylbenzyloxy)ethylamino)-ethyl]-4-methylbenzene sulphonamide(chloro)ruthenium(II), Chloro[(S,S)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulphonamide]ruthenium(II)

A182857-100MG A182857-1G A182857-250MG
77248-320EA 25 USD
77248-320 77248-322 77248-324
(S,S)-Ts-DENEB® 98%
(S,S)-Ts-DENEB®

Precaución: for Research Use Only

Formula: C₃₁H₃₃ClN₂O₃RuS
MDL Number: MFCD22666040
CAS Number: 1384974-37-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR