4-Bromomethyl-1,2-dihydroquinoline-2-one 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Bromomethyl)quinolin-2(1H)-one

A182795-100G A182795-10G A182795-25G A182795-500G A182795-5G
77287-314EA 38.11 USD
77287-314 77287-316 77287-318 77287-320 77287-322
4-Bromomethyl-1,2-dihydroquinoline-2-one 97%
4-Bromomethyl-1,2-dihydroquinoline-2-one

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
MDL Number: MFCD03426152
CAS Number: 4876-10-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR