(R)-(-)-Nipecotic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Piperidine-3-carboxylic acid, (R)-(-)-3-Piperidinecarboxylic acid, D-Nipecotic acid

A182750-1G A182750-250MG A182750-25G A182750-5G A182750-100G
77287-306EA 10.3 USD
77287-306 77287-308 77287-310 77287-312 77748-191
(R)-(-)-Nipecotic acid 98%
(R)-(-)-Nipecotic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
MDL Number: MFCD01630787
CAS Number: 25137-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR