(S)-2-Aminopropanamide hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Ala-NH2.HCl, (S)-2-Aminopropionamide Hydrochloride, L-Alanamine Hydrochloride, H-Ala-NH₂·HCl, L-Alaninamide hydrochloride
A182679-1000G
A182679-100G
A182679-10G
A182679-25G
A182679-500G
A182679-5G
77287-294EA
326.99
USD
77287-294
77287-296
77287-298
77287-300
77287-302
77287-304
(S)-2-Aminopropanamide hydrochloride 98%
(S)-2-Aminopropanamide hydrochloride
Precaución: for Research Use Only
Formula:
C₃H₉ClN₂O MW: 124.57 g/mol |
MDL Number:
MFCD00066145 CAS Number: 33208-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...