1-Bromo-2-fluoro-3-nitrobenzene 98%

Supplier: Ambeed
Warning

A182658-100G A182658-10G A182658-1G A182658-25G A182658-500G A182658-5G
77274-148EA 261 USD
77274-148 77274-150 77274-152 77274-154 77274-156 77274-158
1-Bromo-2-fluoro-3-nitrobenzene 98%
1-Bromo-2-fluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
MDL Number: MFCD05663754
CAS Number: 58534-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR