4-(4-Fluorobenzyl)piperidine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A182620-1G
A182620-250MG
A182620-5G
77287-288EA
41.2
USD
77287-288
77287-290
77287-292
4-(4-Fluorobenzyl)piperidine hydrochloride 97%
4-(4-Fluorobenzyl)piperidine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₂H₁₆FN·ClH MW: 229.723 g/mol |
MDL Number:
MFCD03840140 CAS Number: 92822-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...