3-(Diethylamino)-1-propanol 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Diethylamino)propan-1-ol, 3-Diethylamino-1-propanol

A182533-100G A182533-10G A182533-1G A182533-25G A182533-5G
77274-122EA 114 USD
77274-122 77274-124 77274-126 77274-128 77274-130
3-(Diethylamino)-1-propanol 97%
3-(Diethylamino)-1-propanol

Precaución: for Research Use Only

Formula: C₇H₁₇NO
MW: 131.22 g/mol
MDL Number: MFCD00002947
CAS Number: 622-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR